Learn More
Cole Parmer Cole-Parmer Sanitary Clamp, 304L SS, 1" - 1.5" DFS Item
Manufacturer:  Cole Parmer 3080075
Easily connects pipes or fittings of different types and is ideal for pharmaceutical needs. Meets FDA requirements and 3A sanitary standards. Compatible with standard 316L or 304 adapters. Can be used with corrosive acids and bases.
Catalog No. NC0202204