Learn More
Diagnostic Test Group CLARITY MDMA CASS BX25
SDP
Supplier:  Diagnostic Test Group DTGMDMAC
Encompass_Preferred
Clarity MDMA/Ecstasy Cassette 500ng/mL in.CLIA Waived in.
Catalog No. 50-143-102
Explore available promotions