Learn More
CLASP2 Monoclonal Antibody (KT68), Invitrogen™
Rat Monoclonal Antibody
Supplier:  Thermo Scientific MA547709
Encompass
Catalog No. PIMA547709
Add to Cart
Description
Description
The antibody detects a 160 kDa* doublet corresponding to the molecular mass of CLASP2 on SDS-PAGE immunoblots of human HeLa, mouse brain, and rat PC12 cells. The antibody also detects CLASP2 by immunocytochemistry in HeLa and A431 cells.
CLASP2 Monoclonal antibody specifically detects CLASP2 in Human, Mouse, Rat samples. It is validated for Immunocytochemistry, Western Blot
TRUSTED_SUSTAINABILITY
Specifications
Specifications
CLASP2
Monoclonal
∽1.0 mg/mL
PBS with 0.05% sodium azide
P49615, Q00535, Q03114
Rat
Protein G
RUO
1020, 12568, 140908
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
IgG2b
Immunocytochemistry, Western Blot
KT68
Unconjugated
CDK5
1500004F14Rik; 8030404L10Rik; C77448; CLASP2; CLASP2alpha; CLASP2beta; CLASP2gamma; CLIP associating protein 2; CLIP-associating protein 2; CLIP-associating protein CLASP2; cytoplasmic linker associated protein 2; cytoplasmic linker-associated protein 2; hOrbit2; Kiaa0627; mKIAA0627; multiple asters (Mast)-like homolog 2; protein Orbit homolog 2
Clones DC17+DC34 were generated from a recombinant full-length human Cdk5 sequence.
100 μL
Primary
Human, Mouse, Rat
Liquid
Recommendations
Recommendations