Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Claspin Rabbit anti-Human, Polyclonal, Proteintech Non-distribution product as customer accommodation.

Rabbit Polyclonal Antibody

Manufacturer:  Proteintech Group IncSupplier Diversity Partner 232061AP

Catalog No. 50-173-4020


Add to cart

Description

Description

Xenopus claspin is an essential upstream regulator of checkpoint kinase 1 and triggers a checkpoint arrest of the cell cycle in the presence of DNA templates in Xenopus egg extracts. The human gene appears to be the homolog Xenopus claspin and its function has not been determined.
Specifications

Specifications

Claspin
0.29 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Q9HAW4
CLASPIN, CLSPN, hClaspin
Rabbit
IgG
150 μL
-20°C
Primary
63967
Immunocytochemistry, Immunofluorescence, Immunoprecipitation, Western Blot
Unconjugated
CLSPN
Liquid
CLSPN
Claspin Fusion Protein Ag19637
Antigen Affinity Chromatography
RUO
Antibody
Polyclonal
Human
Documents
Provide Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit