Learn More
National Safety Apparel™ ArcGuard™ Class 0 Rubber Voltage Gloves Kit
Kit includes a pair rubber electrical insulating gloves, a pair of 10 in. leather glove protectors, and a convenient storage bag. Class 0 insulating gloves will protect workers from up to 1000 V.
$242.01 - $282.54
Specifications
Certifications/Compliance | ANSI/ASTM D120, Class 0, Type I, NFPA 70E |
---|---|
Glove Cuff Style | Rolled |
Length (English) | 11 in. |
Material | Rubber, Leather |
Product Type | Electrical Insulating Gloves Kit |
Catalog Number | Mfr. No. | Color | Size | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Color | Size | Price | Quantity & Availability | ||||
17-355-015
![]() |
National Safety Apparel™
KITGC0B08 |
Black | 8 |
Each for $242.01
|
|
17-355-016
![]() |
National Safety Apparel™
KITGC0B09 |
Black | 9 |
Each for $282.54
|
|
17-355-017
![]() |
National Safety Apparel™
KITGC0B10 |
Black | 10 |
Each for $282.54
|
|
17-355-018
![]() |
National Safety Apparel™
KITGC0B11 |
Black | 11 |
Each for $282.54
|
|
17-355-019
![]() |
National Safety Apparel™
KITGC0B12 |
Black | 12 |
Each for $261.17
|
|
17-355-020
![]() |
National Safety Apparel™
KITGC0R08 |
Red | 8 |
Each for $282.54
|
|
17-355-021
![]() |
National Safety Apparel™
KITGC0R09 |
Red | 9 |
Each for $282.54
|
|
17-355-022
![]() |
National Safety Apparel™
KITGC0R10 |
Red | 10 |
Each for $282.54
|
|
17-355-023
![]() |
National Safety Apparel™
KITGC0R11 |
Red | 11 |
Each for $282.54
|
|
17-355-024
![]() |
National Safety Apparel™
KITGC0R12 |
Red | 12 |
Each for $255.17
|
|
17-355-025
![]() |
National Safety Apparel™
KITCG0Y08 |
Yellow | 8 |
Each for $273.77
|
|
17-355-026
![]() |
National Safety Apparel™
KITCG0Y09 |
Yellow | 9 |
Each for $242.01
|
|
17-355-027
![]() |
National Safety Apparel™
KITCG0Y10 |
Yellow | 10 |
Each for $252.42
|
|
17-355-028
![]() |
National Safety Apparel™
KITCG0Y11 |
Yellow | 11 |
Each for $252.42
|
|
17-355-029
![]() |
National Safety Apparel™
KITCG0Y12 |
Yellow | 12 |
Each for $252.42
|
|
Description
- Electrical insulating gloves protect against electrical shock and other arc flash hazards
- Wear with included leather glove protectors for additional resistance against mechanical hazards
- Compliant with ANSI/ASTM D120 and NFPA 70E standards
- Class 0 gloves offer protection from up to 1000 V
- Gloves should be used within one year of marked test date
- When not in use, lay flat in the included protective bag
- Store away from direct sunlight, high heat, and humidity
Specifications
ANSI/ASTM D120, Class 0, Type I, NFPA 70E | |
11 in. | |
Electrical Insulating Gloves Kit | |
1000 V |
Rolled | |
Rubber, Leather | |
KITGC0 |