Learn More
National Safety Apparel™ ArcGuard™ Class 0 Rubber Voltage Gloves
Gloves are constructed of natural rubber and compliant with ANSI/ASTM D120 and NFPA 70E standards for use around electrical hazards. Class 0 insulating gloves will protect workers from up to 1000 V.
$136.63 - $415.50
Specifications
Certifications/Compliance | ANSI/ASTM D120, Class 0, Type I, NFPA 70E-2015 |
---|---|
Glove Cuff Style | Rolled |
Length (English) | 11 in. |
Material | Rubber |
Series | GC0B |
Catalog Number | Mfr. No. | Color | Size | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Color | Size | Price | Quantity & Availability | ||||
17-355-232
![]() |
National Safety Apparel™
GC0B08 |
Black | 8 |
Pair for $415.50
|
|
17-355-233
![]() |
National Safety Apparel™
GC0B09 |
Black | 9 |
Pair for $415.50
|
|
17-355-234
![]() |
National Safety Apparel™
GC0B10 |
Black | 10 |
Pair for $415.50
|
|
17-355-235
![]() |
National Safety Apparel™
GC0B11 |
Black | 11 |
Pair for $415.50
|
|
17-355-236
![]() |
National Safety Apparel™
GC0B12 |
Black | 12 |
Pair for $415.50
|
|
17-355-242
![]() |
National Safety Apparel™
GC0R08 |
Red | 8 |
Pair for $136.63
|
|
17-355-243
![]() |
National Safety Apparel™
GC0R09 |
Red | 9 |
Pair for $136.63
|
|
17-355-244
![]() |
National Safety Apparel™
GC0R10 |
Red | 10 |
Pair for $415.50
|
|
17-355-245
![]() |
National Safety Apparel™
GC0R11 |
Red | 11 |
Pair for $371.81
|
|
17-355-246
![]() |
National Safety Apparel™
GC0R12 |
Red | 12 |
Pair for $369.53
|
|
17-355-237
![]() |
National Safety Apparel™
GC0Y08 |
Yellow | 8 |
Pair for $415.50
|
|
17-355-238
![]() |
National Safety Apparel™
GC0Y09 |
Yellow | 9 |
Pair for $415.50
|
|
17-355-239
![]() |
National Safety Apparel™
GC0Y10 |
Yellow | 10 |
Pair for $415.50
|
|
17-355-241
![]() |
National Safety Apparel™
GC0Y12 |
Yellow | 12 |
Pair for $415.50
|
|
Description
- Class 0 rubber electrical insulating gloves
- Compliant with ANSI/ASTM D120 and NFPA 70E-2015 standards for use around electrical and arc flash hazards
- Gloves should be paired with 10 in. leather glove protectors for additional protection against mechanical hazards
- Must be used within 12 months of the stamped test date
Specifications
ANSI/ASTM D120, Class 0, Type I, NFPA 70E-2015 | |
11 in. | |
GC0B |
Rolled | |
Rubber | |
1000 V |