Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
National Safety Apparel™ ArcGuard™ Class 00 Rubber Voltage Gloves Kit

Click to view available options
Color:
Black
Red
Yellow
Size:
8
9
10
11
12
Description
- Electrical insulating gloves protect against electrical shock and other arc flash hazards
- Wear with included leather glove protectors for additional resistance against mechanical hazards
- Compliant with ANSI/ASTM D120 and NFPA 70E standards
- Class 00 gloves offer protection from up to 500 V
- Gloves should be used within one year of marked test date
- When not in use, lay flat in the included protective bag
- Store away from direct sunlight, high heat, and humidity
Specifications
Certifications/Compliance | ANSI/ASTM D120, Class 00, Type I, NFPA 70E-2015 |
Applications | Industrial |
Color | Black |
Material | Rubber, Leather |
Product Type | Electrical Insulating Gloves Kit |
Size | 10 |
For Use With (Application) | Electrical Utility Work, Industrial Plant Maintenance, and Power Generation |
Glove Cuff Style | Straight |
Includes | Natural Rubber Electrical Insulating Gloves, Leather Glove Protectors, and Storage Bag |
Series | KITGC00 |
Show More |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title