Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
National Safety Apparel™ ArcGuard™ Class 00 Rubber Voltage Gloves Kit

Click to view available options
Color:
Black
Red
Yellow
Size:
8
9
10
11
12
Description
- Electrical insulating gloves protect against electrical shock and other arc flash hazards
- Wear with included leather glove protectors for additional resistance against mechanical hazards
- Compliant with ANSI/ASTM D120 and NFPA 70E standards
- Class 00 gloves offer protection from up to 500 V
- Gloves should be used within one year of marked test date
- When not in use, lay flat in the included protective bag
- Store away from direct sunlight, high heat, and humidity
Specifications
Certifications/Compliance | ANSI/ASTM D120, Class 00, Type I, NFPA 70E-2015 |
Applications | Industrial |
Color | Red |
Material | Rubber, Leather |
Product Type | Electrical Insulating Gloves Kit |
Size | 8 |
For Use With (Application) | Electrical Utility Work, Industrial Plant Maintenance, and Power Generation |
Glove Cuff Style | Straight |
Includes | Natural Rubber Electrical Insulating Gloves, Leather Glove Protectors, and Storage Bag |
Series | KITGC00R |
Show More |