Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

National Safety Apparel™ ArcGuard™ Class 00 Rubber Voltage Gloves Kit
SDP

Click to view available options
Color:
Black
Red
Yellow
Size:
8
9
10
11
12
This item is not returnable. View return policy
This item is not returnable. View return policy

Kit includes a pair of rubber electrical insulating gloves, a pair of 10 in. leather glove protectors, and a convenient storage bag. Class 00 insulating gloves will protect workers from up to 500 V.

  • Electrical insulating gloves protect against electrical shock and other arc flash hazards
  • Wear with included leather glove protectors for additional resistance against mechanical hazards
  • Compliant with ANSI/ASTM D120 and NFPA 70E standards
  • Class 00 gloves offer protection from up to 500 V
  • Gloves should be used within one year of marked test date
  • When not in use, lay flat in the included protective bag
  • Store away from direct sunlight, high heat, and humidity
Certifications/Compliance ANSI/ASTM D120, Class 00, Type I, NFPA 70E-2015
Applications Industrial
Color Yellow
Material Rubber, Leather
Product Type Electrical Insulating Gloves Kit
Size 9
For Use With (Application) Electrical Utility Work, Industrial Plant Maintenance, and Power Generation
Glove Cuff Style Straight
Includes Natural Rubber Electrical Insulating Gloves, Leather Glove Protectors, and Storage Bag
Series KITGC00Y
Length (English) 11 in.
Length (Metric) 28 cm
Voltage 500 V
Show More Show Less