Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

National Safety Apparel™ Class 00 ArcGuard™ Rubber Voltage Glove Kit

National Safety Apparel™ Class 00 ArcGuard™ Rubber Voltage Glove Kit includes leather gloves, class 00 rubber voltage gloves, and a glove bag.

$264.00

Specifications

Certifications/Compliance ASTM CLASS 00
Material Leather, Rubber
For Use With (Application) Voltage, electrical, arc flash
View More Specs

Products 15
Catalog Number Mfr. No. Color Size Price Quantity & Availability  
Catalog Number Mfr. No. Color Size Price Quantity & Availability  
17-355-000
View Documents
National Safety Apparel™
KITGC00B08
Black 8 Pair for $264.00
Add to cart
 
17-355-001
View Documents
National Safety Apparel™
KITGC00B09
Black 9 Pair for $264.00
Add to cart
 
17-355-002
View Documents
National Safety Apparel™
KITGC00B10
Black 10 Pair for $264.00
Add to cart
 
17-355-003
View Documents
National Safety Apparel™
KITGC00B11
Black 11 Pair for $264.00
Add to cart
 
17-355-004
View Documents
National Safety Apparel™
KITGC00B12
Black 12 Pair for $264.00
Add to cart
 
17-355-005
View Documents
National Safety Apparel™
KITGC00R08
Red 8 Pair for $264.00
Add to cart
 
17-355-006
View Documents
National Safety Apparel™
KITGC00R09
Red 9 Pair for $264.00
Add to cart
 
17-355-007
View Documents
National Safety Apparel™
KITGC00R10
Red 10 Pair for $264.00
Add to cart
 
17-355-008
View Documents
National Safety Apparel™
KITGC00R11
Red 11 Pair for $264.00
Add to cart
 
17-355-009
View Documents
National Safety Apparel™
KITGC00R12
Red 12 Pair for $264.00
Add to cart
 
17-355-010
View Documents
National Safety Apparel™
KITGC00Y08
Yellow 8 Pair for $264.00
Add to cart
 
17-355-011
View Documents
National Safety Apparel™
KITGC00Y09
Yellow 9 Pair for $264.00
Add to cart
 
17-355-012
View Documents
National Safety Apparel™
KITGC00Y10
Yellow 10 Pair for $264.00
Add to cart
 
17-355-013
View Documents
National Safety Apparel™
KITGC00Y11
Yellow 11 Pair for $264.00
Add to cart
 
17-355-014
View Documents
National Safety Apparel™
KITGC00Y12
Yellow 12 Pair for $264.00
Add to cart
 
Description

Description

  • Fabric/Material: Rubber, Leather
  • Rubber voltage gloves are electrically tested and marked with initial test date
  • Class 00 - Max Use Voltage 500 V AC, 750 V DC
  • Imported Includes: Class 00 Rubber Voltage Glove Kit Includes:
  • 10 in. Leather Protectors
  • Glove Bag
Specifications

Specifications

ASTM CLASS 00
Leather, Rubber
Voltage, electrical, arc flash
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit