Learn More
Honeywell Salisbury™ Class 00 Blue Salcor™ Synthetic Rubber Lineman Gloves Kit
Kit includes a pair of type II Salcor™ synthetic rubber electrical insulating gloves, a pair of compatible leather glove protectors, and a storage bag. Class 00 insulating gloves will protect workers from up to 500 V.
$109.00 - $115.00
Specifications
Certifications/Compliance ANSI/ASTM D120, Class 00, Type II, ASTM F696
Color Blue
Glove Cuff Style Rolled
Includes Salcor™ Rubber Electrical Insulating Gloves, Leather Glove Protectors, and Storage Bag
Material Salcor™ Synthetic Rubber
View More Specs
Products 12
Catalog Number Mfr. No. Size Length (English) Price Quantity & Availability  
Catalog Number Mfr. No. Size Length (English) Price Quantity & Availability  
19-139-710
View Documents
Honeywell Salisbury™
GK0011BL/7
7 11 in.
Each for $109.00
Add to cart
19-139-734
View Documents
Honeywell Salisbury™
GK0014BL/7
7 14 in.
Each for $115.00
Add to cart
19-139-711
View Documents
Honeywell Salisbury™
GK0011BL/8
8 11 in.
Each for $109.00
Add to cart
19-139-735
View Documents
Honeywell Salisbury™
GK0014BL/8
8 14 in.
Each for $115.00
Add to cart
19-139-712
View Documents
Honeywell Salisbury™
GK0011BL/9
9 11 in.
Each for $109.00
Add to cart
19-139-736
View Documents
Honeywell Salisbury™
GK0014BL/9
9 14 in.
Each for $115.00
Add to cart
19-139-713
View Documents
Honeywell Salisbury™
GK0011BL/10
10 11 in.
Each for $109.00
Add to cart
19-139-737
View Documents
Honeywell Salisbury™
GK0014BL/10
10 14 in.
Each for $115.00
Add to cart
19-139-714
View Documents
Honeywell Salisbury™
GK0011BL/11
11 11 in.
Each for $109.00
Add to cart
19-139-738
View Documents
Honeywell Salisbury™
GK0014BL/11
11 14 in.
Each for $115.00
Add to cart
19-139-715
View Documents
Honeywell Salisbury™
GK0011BL/12
12 11 in.
Each for $109.00
Add to cart
19-139-739
View Documents
Honeywell Salisbury™
GK0014BL/12
12 14 in.
Each for $115.00
Add to cart
Description
Description
  • Glove kit includes a pair of Type II Salcor™ synthetic rubber electrical insulating gloves, and a pair of compatible leather protector gloves
  • Leather protectors must be no less than 0.5 in. shorter than the cuff of the insulating glove
  • Voltage Class 00 gloves will protect workers up to 500 V
  • Type II Salcor™ synthetic rubber is ozone resistant
  • Canvas bag has a wide opening for easy insertion
Compliance
ANSI/ASTM D120, Class 00, Type II
Specifications
Specifications
ANSI/ASTM D120, Class 00, Type II, ASTM F696
Rolled
Salcor™ Synthetic Rubber
500 V
Blue
Salcor™ Rubber Electrical Insulating Gloves, Leather Glove Protectors, and Storage Bag
GK0011BL