Learn More
Honeywell Salisbury™ Class 00 Natural Rubber Lineman Gloves Kit - 11 in.
Kit includes a pair of natural rubber electrical insulating gloves, a pair of compatible leather glove protectors, and a storage bag. Class 00 insulating gloves will protect workers from up to 500 V.
$109.00 - $112.00
Specifications
Certifications/Compliance ANSI/ASTM D120, Class 00, Type I, ASTM F696
Glove Cuff Style Rolled
Includes Natural Rubber Electrical Insulating Gloves, Leather Glove Protectors, and a Storage Bag
Length (English) 11 in.
Material Rubber
View More Specs
Products 17
Catalog Number Mfr. No. Color Size Price Quantity & Availability  
Catalog Number Mfr. No. Color Size Price Quantity & Availability  
19-139-723
View Documents
Honeywell Salisbury™
GK0011B/8
Black 8
Each for $109.00
Add to cart
19-139-724
missing translation for 'viewDocuments'
Honeywell Salisbury™
GK0011B/9
Black 9
Each for $109.00
Add to cart
19-139-725
missing translation for 'viewDocuments'
Honeywell Salisbury™
GK0011B/10
Black 10
Each for $109.00
Add to cart
19-139-727
missing translation for 'viewDocuments'
Honeywell Salisbury™
GK0011B/12
Black 12
Each for $109.00
Add to cart
19-139-704
missing translation for 'viewDocuments'
Honeywell Salisbury™
GK0011R/7
Red 7
Each for $109.00
Add to cart
19-139-705
missing translation for 'viewDocuments'
Honeywell Salisbury™
GK0011R/8
Red 8
Each for $109.00
Add to cart
19-139-706
missing translation for 'viewDocuments'
Honeywell Salisbury™
GK0011R/9
Red 9
Each for $109.00
Add to cart
19-139-707
missing translation for 'viewDocuments'
Honeywell Salisbury™
GK0011R/10
Red 10
Each for $109.00
Add to cart
19-139-708
missing translation for 'viewDocuments'
Honeywell Salisbury™
GK0011R/11
Red 11
Each for $109.00
Add to cart
19-139-709
missing translation for 'viewDocuments'
Honeywell Salisbury™
GK0011R/12
Red 12
Each for $109.00
Add to cart
19-139-716
missing translation for 'viewDocuments'
Honeywell Salisbury™
GK0011Y/7
Yellow 7
Each for $112.00
Add to cart
19-139-717
missing translation for 'viewDocuments'
Honeywell Salisbury™
GK0011Y/8
Yellow 8
Each for $112.00
Add to cart
19-139-718
missing translation for 'viewDocuments'
Honeywell Salisbury™
GK0011Y/9
Yellow 9
Each for $112.00
Add to cart
19-139-719
missing translation for 'viewDocuments'
Honeywell Salisbury™
GK0011Y/10
Yellow 10
Each for $112.00
Add to cart
19-139-720
missing translation for 'viewDocuments'
Honeywell Salisbury™
GK0011Y/11
Yellow 11
Each for $112.00
Add to cart
19-139-722
missing translation for 'viewDocuments'
Honeywell Salisbury™
GK0011B/7
Black 7 This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. N/A
19-139-726
missing translation for 'viewDocuments'
Honeywell Salisbury™
GK0011B/11
Black 11 This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. N/A
Description
Description
  • Glove kit includes a pair of natural rubber electrical insulating gloves, and a pair of compatible leather protector gloves
  • Leather protectors must be no less than 0.5 in. shorter than the cuff of the insulating glove
  • Voltage Class 00 gloves will protect workers up to 500 V
  • Canvas bag has a wide opening for easy insertion
Compliance
ANSI/ASTM D120, Class 00, Type I
Specifications
Specifications
ANSI/ASTM D120, Class 00, Type I, ASTM F696
Natural Rubber Electrical Insulating Gloves, Leather Glove Protectors, and a Storage Bag
Rubber
500 V
Rolled
11 in.
GK0011B