Learn More
Fisherbrand™ Class 10 Clean Room/ESD Vinyl Chair - Desk Height with Medium Back and ESD Casters in ESD Vinyl
Fisherbrand™ Class 10 Clean Room/ESD Vinyl Chairs with Medium Back are height adjustable for 31 in. work surfaces and feature clean room and electro-static dissipating components such as vinyl and self-braking casters and larger backs. Ideal for static sensitive environments.
$555.00
Specifications
Adjustable Yes
Footrest No
With Armrest No
With Backrest Yes
With Casters Yes
View More Specs
Products 2
Catalog Number Color Price Quantity & Availability  
Catalog Number Color Price Quantity & Availability  
14-000-495
View Documents
Blue
Each for $555.00
Add to cart
14-000-496
View Documents
Black
Each for $555.00
Add to cart
Description
Description
  • Medium Back
  • Adjustable from 18.5-23.75 inches from the top of the seat to the floor
  • Features manual back height and back depth adjustments
  • Electro-static dissipating components to ensure proper grounding
  • Cast-aluminum base
  • Dual-wheel, self-braking, non-marring ESD casters suitable for hard floors
  • Electro-static dissipating vinyl
  • Class 10/ISO 4 Clean Room
  • Meets ANSI/ESD STM12.1-2013
Specifications
Specifications
Yes
No
Yes
Class 10/ISO Class 4
No
Yes
No