Learn More
Fisherbrand™ Class 10 Clean Room/ESD Vinyl Chair - Medium Bench Height with Chrome Foot Ring and ESD Casters in ESD Vinyl
SCI_ED
Fisherbrand™ Class 10 Clean Room/ESD Vinyl Chairs are height adjustable for 36 in. work surfaces and feature clean room and electro-static dissipating components such as vinyl and self-braking casters & chrome adjustable foot ring. Ideal for static sensitive environments.
 View more versions of this product
Catalog No. S36589
Add to cart
Description
Description
  • Adjustable from 21.5-29 inches from the top of the seat to the floor
  • Features manual back height and back depth adjustments
  • Electro-static dissipating components to ensure proper grounding
  • Chrome adjustable foot ring
  • Cast-aluminum base
  • Dual-wheel, self-braking, non-marring ESD casters suitable for hard floors
  • Electro-static dissipating vinyl
  • Class 10/ISO 4 Clean Room
  • Meets ANSI/ESD STM12.1-2013
Specifications
Specifications
Standard
Class 10/ISO Class 4
Electro-Static Dissipating, dual-wheel, non-marring casters (suitable for both hard and carpeted floors)
20 in.
ESD Upholstery
No
Yes
21.5 to 29 in.
Polished Cast Aluminum
136 kg
Yes
Blue
Yes
Round tube chrome foot ring with internal lock
Medium Bench Height
Yes
No
300 lb.
Cleanroom ESD Chair