Learn More
Fisherbrand™ Class 10 Clean Room/ESD Vinyl Chair - Medium Bench Height with Medium Back, Chrome Foot Ring, and ESD Casters in ESD Vinyl
Fisherbrand™ Class 10 Clean Room/ESD Vinyl Chairs with Medium Back are height adjustable for 36 in. work surfaces & feature clean room & electro-static dissipating components such as vinyl and self-braking casters, larger backs, & chrome adjustable foot rings. Ideal for static sensitive environments.
$599.50
Specifications
Adjustable Yes
Footrest Yes
With Armrest No
With Backrest Yes
With Casters Yes
View More Specs
Products 2
Catalog Number Color Price Quantity & Availability  
Catalog Number Color Price Quantity & Availability  
14-000-481
View Documents
Blue
Each for $599.50
Add to cart
14-000-482
View Documents
Black
Each for $599.50
Add to cart
Description
Description
  • Medium Back
  • Adjustable from 21.5-29 inches from the top of the seat to the floor
  • Features manual back height and back depth adjustments
  • Electro-static dissipating components to ensure proper grounding
  • Chrome adjustable foot ring
  • Cast-aluminum base
  • Dual-wheel, self-braking, non-marring ESD casters suitable for hard floors
  • Electro-static dissipating vinyl
  • Class 10/ISO 4 Clean Room
  • Meets ANSI/ESD STM12.1-2013
Specifications
Specifications
Yes
No
Yes
Class 10/ISO Class 4
Yes
Yes
No