Learn More
Fisherbrand™ Class 10 Clean Room Vinyl Chair Chrome - Desk Height with Casters in Grade A Vinyl
Fisherbrand™ Class 10 Clean Room Chairs are height adjustable for 31 in. work surfaces and feature clean room components and self-braking casters. Grade A vinyl, in a variety of colors, offer additional antibacterial properties and a high-end look and feel.
$665.00
Specifications
Base Material Polished Cast Aluminum
Foot Type Chrome-hooded dual-wheel, self-braking, non-marring casters (suitable for both hard and carpeted floors)
Footrest No
Material Vinyl
Weight (English) 31 lb.
View More Specs
Products 6
Catalog Number Color Price Quantity & Availability  
Catalog Number Color Price Quantity & Availability  
15-001-738
View Documents
Marine Blue
Each for $665.00
Add to cart
15-001-751
View Documents
Taupe
Each for $665.00
Add to cart
15-001-764
View Documents
Borscht
Each for $665.00
Add to cart
15-001-777
View Documents
Storm
Each for $665.00
Add to cart
15-001-790
View Documents
Carbon
Each for $665.00
Add to cart
15-001-803
View Documents
Sterling
Each for $665.00
Add to cart
Description
Description
  • Adjustable from 18.5 to 23.75 inches from the top of the seat to the floor
  • Features manual back height and back depth adjustments
  • Cast-aluminum base
  • Chrome-hooded, dual-wheel, self-braking, non-marring casters suitable for hard floors
  • Antifungal/antibacterial Grade A vinyl
  • Additional antibacterial protections
  • High end look and feel
  • Variety of color options to suit any work space
  • Class 10/ISO 4 Clean Room
Specifications
Specifications
Polished Cast Aluminum
No
31 lb.
Yes
Chrome-hooded dual-wheel, self-braking, non-marring casters (suitable for both hard and carpeted floors)
Vinyl
No
No