Learn More
Fisherbrand™ Class 10 Clean Room Vinyl Chair Chrome - Medium Bench Height with Chrome Foot Ring and Casters in Grade B Vinyl Products for Science Education
Fisherbrand™ Class 10 Clean Room Chairs are height adjustable for 36 in. work surfaces and feature clean room components, chrome adjustable foot ring, and self-braking casters. Antibacterial/Antifungal Grade B vinyl is easily cleaned making it ideal for clean room environments.
$325.00
Specifications
Adjustable Yes
Base Material Polished Cast Aluminum
Material Vinyl
Footrest Yes
With Armrest No
View More Specs
Products 4
Catalog Number Color Price Quantity & Availability  
Catalog Number Color Price Quantity & Availability  
S0100900 Products for Science Education
View Documents
Taupe
Each for $325.00
Add to cart
S0100902 Products for Science Education
View Documents
Dove
Each for $325.00
Add to cart
S0100912 Products for Science Education
View Documents
White
Each for $325.00
Add to cart
S0100938 Products for Science Education
View Documents
Orange
Each for $325.00
Add to cart
Description
Description
  • Adjustable from 21.5-29 inches from the top of the seat to the floor
  • Features manual back height and back depth adjustments
  • 20-Inch Chrome adjustable foot ring with internal lock
  • Cast-aluminum base
  • Chrome-hooded, dual-wheel, self-braking, non-marring casters suitable for hard floors
  • Antifungal/antibacterial Grade B vinyl
  • Variety of color options to suit any work space
  • Class 10/ISO 4 Clean Room
Specifications
Specifications
Yes
Vinyl
No
Yes
Medium Bench Height
Polished Cast Aluminum
Yes
Yes
No
Class 10/ISO Class 4