Learn More
Fisherbrand™ Class 10 Erie Poly Clean Room Chair - High Bench Height with Adjustable Arms, Chrome Foot Ring and Stationary Glides in Poly
SCI_ED
Fisherbrand™ Class 10 Erie Clean Room Chairs are height adjustable for 42 in. work surfaces and feature clean room components, adjustable arms, and stationary glides. Self-skinning polyurethane is chemical, soil, and puncture resistant and easily cleaned making it ideal for clean room room environments.
 View more versions of this product
Catalog No. S41435
Add to cart
Description
Description
  • Adjustable from 23.5-33.5 inches from the top of the seat to the floor
  • Features manual back height and back depth adjustments
  • Built in handle for ease of movement
  • 2-way adjustable arm rests adjust up and down
  • 20-Inch Chrome adjustable foot ring with internal lock
  • Cast-aluminum base
  • Stationary, non-marring glides suitable for hard floors
  • Chemical, soil, and puncture resistant polyurethane
  • Class 100/ISO 5 Clean Room
Specifications
Specifications
Standard
2-way adjustable (move up and down)
Blue
Yes
Round tube chrome foot ring with internal lock
High Bench Height
Yes
No
23.5 to 33.5 in.
Polished Cast Aluminum
136 kg
Yes
Class 10/ISO Class 4
Black-non-marring bell glides (suitable for both hard and carpeted floors)
20 in.
Polyurethane
46 lb.
Yes
No
300 lb.
Cleanroom Chair