Learn More
Fisherbrand™ Class 10 Erie Polyurethane Clean Room Chair - High Bench Height with Chrome Foot Ring and Stationary Glides in Polyurethane
Fisherbrand™ Class 10 Erie Clean Room Chairs are height adjustable for 42 in. work surfaces and feature clean room components and stationary glides. Self-skinning polyurethane is chemical, soil, and puncture resistant and easily cleaned making it ideal for clean room room environments.
 View more versions of this product
Catalog No. 14-000-523
Add to cart
Description
Description
  • Adjustable from 23.5-33.5 inches from the top of the seat to the floor
  • Features manual back height and back depth adjustments
  • Built in handle for ease of movement
  • 20-Inch Chrome adjustable foot ring with internal lock
  • Cast-aluminum base
  • Stationary, non-marring glides suitable for hard floors
  • Chemical, soil, and puncture resistant polyurethane
  • Class 10/ISO 4 Clean Room
Specifications
Specifications
Standard
Class 10/ISO Class 4
Black-non-marring bell glides (suitable for both hard and carpeted floors)
20 in.
Polyurethane
36 lb.
Yes
No
300 lb.
Cleanroom Chair
Yes
Black
Yes
Round tube chrome foot ring with internal lock
High Bench Height
No
No
23.5 to 33.5 in.
Polished Cast Aluminum
136 kg