Learn More
Fisherbrand™ Class 10 Erie Polyurethane Clean Room Chair - High Bench Height with Chrome Foot Ring and Stationary Glides in Polyurethane
Fisherbrand™ Class 10 Erie Clean Room Chairs are height adjustable for 42 in. work surfaces and feature clean room components and stationary glides. Self-skinning polyurethane is chemical, soil, and puncture resistant and easily cleaned making it ideal for clean room room environments.
$336.50 - $357.50
Specifications
Adjustable Yes
Material Polyurethane
Footrest Yes
With Armrest No
With Backrest Yes
View More Specs
Products 2
Catalog Number Color Price Quantity & Availability  
Catalog Number Color Price Quantity & Availability  
14-000-821
View Documents
Blue
Each for $357.50
Add to cart
14-000-523
View Documents
Black
Each for $336.50
Add to cart
Description
Description
  • Adjustable from 23.5-33.5 inches from the top of the seat to the floor
  • Features manual back height and back depth adjustments
  • Built in handle for ease of movement
  • 20-Inch Chrome adjustable foot ring with internal lock
  • Cast-aluminum base
  • Stationary, non-marring glides suitable for hard floors
  • Chemical, soil, and puncture resistant polyurethane
  • Class 10/ISO 4 Clean Room
Specifications
Specifications
Yes
Yes
Yes
No
Polyurethane
No
No
Class 10/ISO Class 4