Learn More
Fisherbrand™ Class 10 Erie Polyurethane Clean Room Chair - Medium Bench Height with Casters in Polyurethane Products for Science Education
Fisherbrand™ Class 10 Erie Clean Room Chairs are height adjustable for 36 in. work surfaces and feature clean room components, adjustable arms, and self-braking casters. Self-skinning polyurethane is chemical, soil, and puncture resistant and easily cleaned making it ideal for clean room room environments.
$246.00 - $262.00
Specifications
Adjustable Yes
Material Polyurethane
Footrest No
With Armrest No
With Backrest Yes
View More Specs
Products 2
Catalog Number Color Price Quantity & Availability  
Catalog Number Color Price Quantity & Availability  
S41440 Products for Science Education
View Documents
Blue
Each for $262.00
Add to cart
S40372 Products for Science Education
View Documents
Black
Each for $246.00
Add to cart
Description
Description
  • Adjustable from 19.5-27 inches from the top of the seat to the floor
  • Features manual back height and back depth adjustments
  • Built in handle for ease of movement
  • Cast-aluminum base
  • Chrome-hooded, dual-wheel, self-braking, non-marring casters suitable for hard floors
  • Chemical, soil, and puncture resistant polyurethane
  • Class 10/ISO 4 Clean Room
Specifications
Specifications
Yes
No
Yes
No
Polyurethane
No
Yes
Class 10/ISO Class 4