Learn More
Fisherbrand™ Class 10 Newport Industrial Polyurethane Clean Room Chair - High Bench Height with Chrome Foot Ring and Casters in Black Polyurethane Products for Science Education
Fisherbrand™ Newport Class 100 Clean Room Chairs are height adjustable for 42 in. work surfaces and feature ergonomic adjustments, clean room components, and self-braking casters. Self-skinning polyurethane is chemical, soil and puncture resistant and easily cleaned making it ideal for clean room environments.
$314.00
Specifications
Adjustable Yes (Pneumatic)
Material Polyurethane
Footrest Yes
With Armrest No
With Backrest Yes
View More Specs
Products 1
Catalog Number Color Price Quantity & Availability  
Catalog Number Color Price Quantity & Availability  
S40357 Products for Science Education
View Documents
Black
Each for $314.00
Add to cart
Description
Description
  • Adjustable from 24.75-35 inches from the top of the seat to the floor
  • Features ergonomic adjustments including seat height
  • Locking back tilt
  • Built in handle for ease of movement
  • 20-Inch Chrome adjustable foot ring with internal lock
  • Cast-aluminum base
  • Chrome-hooded, dual-wheel, self-braking, non-marring casters suitable for hard floors
  • Chemical, soil, and puncture resistant polyurethane
  • Class 10/ISO 4 Clean Room
Specifications
Specifications
Yes (Pneumatic)
Yes
Yes
No
Polyurethane
No
Yes
Class 10/ISO Class 4