Learn More
Fisherbrand™ Class 10 Polyurethane Clean Room Chair - Medium Bench Height with Medium Back and Casters in Black Polyurethane Products for Science Education
Fisherbrand™ Poly Clean Room Chairs with Medium Back are height adjustable for 36 in. work surfaces and feature ergonomic adjustments and self-braking casters. Self-skinning polyurethane is chemical, soil, and puncture resistant and easily cleaned making it ideal for clean room environments.
$306.00 - $342.00
Specifications
Adjustable Yes
Material Polyurethane
Footrest No
With Armrest No
With Backrest Yes
View More Specs
Products 2
Catalog Number Color Price Quantity & Availability  
Catalog Number Color Price Quantity & Availability  
S41217 Products for Science Education
View Documents
Blue
Each for $306.00
Add to cart
S36368 Products for Science Education
View Documents
Black
Each for $342.00
Add to cart
Description
Description
  • Medium Back
  • Adjustable from 20.25-28 inches from the top of the seat to the floor
  • Features ergonomic adjustments including seat height
  • Locking back tilt
  • Cast-aluminum base
  • Chrome-hooded, dual-wheel, self-braking, non-marring casters suitable for hard floors
  • Chemical, soil, and puncture resistant polyurethane
  • Class 100/ISO Class 5 Clean Room
Specifications
Specifications
Yes
No
Yes
No
Polyurethane
No
Yes
Class 10/ISO Class 4