Learn More
Fisherbrand™ Class 10 Polyurethane Clean Room Chair - Medium Bench Height with Medium Back and Stationary Glides in Black Polyurethane
SCI_ED
Fisherbrand™ Poly Clean Room Chairs with Medium Back are height adjustable for 36 in. work surfaces and feature ergonomic adjustments and stationary glides. Self-skinning polyurethane is chemical, soil, and puncture resistant and easily cleaned making it ideal for clean room environments.
 View more versions of this product
Catalog No. S41220
Add to cart
Description
Description
  • Medium Back
  • Adjustable from 20.25-28 inches from the top of the seat to the floor
  • Features ergonomic adjustments including seat height
  • Locking back tilt
  • Cast-aluminum base
  • Stationary, non-marring glides suitable for hard floors
  • Chemical, soil, and puncture resistant polyurethane
  • Class 100/ISO Class 5 Clean Room
Specifications
Specifications
Standard
Class 10/ISO Class 4
Black-non-marring bell glides (suitable for both hard and carpeted floors)
Polyurethane
33 lb.
Yes
No
300 lb.
Cleanroom Chair
Yes
Blue
No
Medium Bench Height
No
No
20.25 to 28 in.
Polished Cast Aluminum
136 kg