Learn More
Fisherbrand™ Class 100 Core Industrial Chrome and Polyurethane Clean Room Chair with Adjustable Arms Products for Science Education
Fisherbrand Class 100 Core Industrial Chrome and Polyurethane Clean Room Chair with Adjustable Arms features industrial grade, puncture, soil, and chemical resistant seat and back. Fully adjustable.
$248.00 - $306.00
Specifications
Arm Style ADJUSTABLE
Base Style CHROME
Capacity 300 lbs
Lift Type PNEUMATIC
Material POLYURETHANE
View More Specs
Products 10
Catalog Number Color Seating Height With Casters Footrest Price Quantity & Availability  
Catalog Number Color Seating Height With Casters Footrest Price Quantity & Availability  
S40364 Products for Science Education View Documents BLACK DESK HEIGHT NO NO Each for $248.00
Add to cart
S40366 Products for Science Education View Documents BLACK HIGH BENCH YES YES Each for $288.00
Add to cart
S40367 Products for Science Education View Documents BLACK HIGH BENCH NO YES Each for $288.00
Add to cart
S40370 Products for Science Education View Documents BLACK MEDIUM BENCH YES NO Each for $282.00
Add to cart
S40371 Products for Science Education View Documents BLACK MEDIUM BENCH YES YES Each for $282.00
Add to cart
S41432 Products for Science Education View Documents BLUE DESK HEIGHT NO NO Each for $264.00
Add to cart
S41434 Products for Science Education View Documents BLUE HIGH BENCH YES YES Each for $306.00
Add to cart
S41435 Products for Science Education View Documents BLUE HIGH BENCH NO YES Each for $306.00
Add to cart
S41438 Products for Science Education View Documents BLUE MEDIUM BENCH YES NO Each for $300.00
Add to cart
S41439 Products for Science Education View Documents BLUE MEDIUM BENCH YES YES Each for $300.00
Add to cart
Description
Description
  • Features ergonomic adjustments
  • 2-way adjustable arms for proper ergonomic arm placement
  • Handle on back pad for ease of movement around the work space
  • Optional 20 in Diameter Chrome Adjustable Foot Ring
  • Suitable for Class 100 (ISO Class 5) Clean Rooms
  • Soil, Puncture, and Chemical Resistant Industrial Grade Polyurethane Seat and Back
  • Non-Marring Casters or Stationary Glides
Certifications
ANSI/BIFMA
Warranty and Services
15-year on parts, lifetime on pneumatic cylinder
Specifications
Specifications
ADJUSTABLE
300 lbs
POLYURETHANE
YES
NO
CHROME
PNEUMATIC
YES
NO