Learn More
Fisherbrand™ Class 100 Newport Industrial Poly Clean Room Chair - High Bench Height with Adj Arms, Chrome Foot Ring, and Stationary Glides in Black Poly Products for Science Education
Fisherbrand™ Newport Class 100 Clean Room Chairs are height adjustable for 42 in. work surfaces and feature ergonomic adjustments, clean room components, arms and glides. Self-skinning polyurethane is chemical, soil and puncture resistant and easily cleaned making it ideal for clean room environments.
$393.00
Specifications
Adjustable Yes (Pneumatic)
Material Polyurethane
Footrest Yes
With Armrest Yes
With Backrest Yes
View More Specs
Products 1
Catalog Number Color Price Quantity & Availability  
Catalog Number Color Price Quantity & Availability  
S40352 Products for Science Education
View Documents
Black
Each for $393.00
Add to cart
Description
Description
  • Adjustable from 24.75-35 inches from the top of the seat to the floor
  • Features ergonomic adjustments including seat height
  • Locking back tilt
  • Built in handle for ease of movement
  • 6-way adjustable arm rests adjust up and down, in and out, and twist toward and away from the user
  • 20-Inch Chrome adjustable foot ring with internal lock
  • Cast-aluminum base
  • Stationary, non-marring glides suitable for hard floors
  • Chemical, soil, and puncture resistant polyurethane
  • Class 100/ISO 5 Clean Room
Specifications
Specifications
Yes (Pneumatic)
Yes
Yes
No
Polyurethane
Yes
No
Class 100/ISO Class 5