Learn More
Fisherbrand™ Class 100 Newport Industrial Poly Clean Room Chair - Medium Bench Height with Adjustable Arms, Chrome Foot Ring, and Casters in Black Poly
Fisherbrand™ Newport Class 100 Clean Room Chairs are height adjustable for 36 in. work surfaces & feature ergonomic adjustments, clean room components, & self-braking casters. Self-skinning polyurethane is chemical, soil & puncture resistant & easily cleaned making it ideal for clean room environments.
$518.50
Specifications
Adjustable Yes (Pneumatic)
Material Polyurethane
Footrest Yes
With Armrest Yes
With Backrest Yes
View More Specs
Products 1
Catalog Number Color Price Quantity & Availability  
Catalog Number Color Price Quantity & Availability  
14-000-514
View Documents
Black
Each for $518.50
Add to cart
Description
Description
  • Adjustable from 20.5-28 inches from the top of the seat to the floor
  • Features ergonomic adjustments including seat height
  • Locking back tilt
  • Built in handle for ease of movement
  • 6-way adjustable arm rests adjust up and down, in and out, and twist toward and away from the user
  • Chrome adjustable foot ring
  • Cast-aluminum base
  • Chrome-hooded, dual-wheel, self-braking, non-marring casters suitable for hard floors
  • Chemical, soil, and puncture resistant polyurethane
  • Class 100/ISO 5 Clean Room
Specifications
Specifications
Yes (Pneumatic)
Yes
Yes
No
Polyurethane
Yes
Yes
Class 100/ISO Class 5