Learn More
Our pricing system is unavailable. You are viewing list price.
Please call 1-800-766-7000 to place your order or try our site again later.
Fisherbrand™ Class 100 Newport Industrial Polyurethane Clean Room Chair - Desk Height with Adjustable Arms and Casters in Black Polyurethane
Fisherbrand™ Newport Class 100 Clean Room Chairs are height adjustable for 31 in. work surfaces & feature ergonomic adjustments, clean room components, arms & casters. Self-skinning polyurethane is chemical, soil & puncture resistant & easily cleaned making it ideal for clean room environments.
Specifications
Adjustable Yes (Pneumatic)
Material Polyurethane
Footrest No
With Armrest Yes
With Backrest Yes
View More Specs
Products 1
Catalog Number Color Price Quantity & Availability  
Catalog Number Color Price Quantity & Availability  
14-000-502
View Documents
Black
Each for $464.00
N/A
Description
Description
  • Adjustable from 17.75-23 inches from the top of the seat to the floor
  • Features ergonomic adjustments including seat height
  • Locking back tilt
  • Built in handle for ease of movement
  • 6-way adjustable arm rests adjust up and down, in and out, and twist toward and away from the user
  • Cast-aluminum base
  • Chrome-hooded, dual-wheel, self-braking, non-marring casters suitable for hard floors
  • Chemical, soil, and puncture resistant polyurethane
  • Class 100/ISO 5 Clean Room
Specifications
Specifications
Yes (Pneumatic)
No
Yes
No
Polyurethane
Yes
Yes
Class 100/ISO Class 5