Learn More
Fisherbrand™ Class 100 Newport Industrial Polyurethane Clean Room Chair - Desk Height with Seat Tilt and Casters in Polyurethane
SCI_ED
Fisherbrand™ Newport Class 100 Clean Room Chairs are height adjustable for 31 in. work surfaces and feature ergonomic adjustments, clean room components, and self-braking casters. Self-skinning polyurethane is chemical, soil and puncture resistant and easily cleaned making it ideal for clean room environments.
 View more versions of this product
Catalog No. S40351
Add to cart
Description
Description
  • Adjustable from 17.75-23 inches from the top of the seat to the floor
  • Features ergonomic adjustments including seat height
  • Locking back tilt
  • Built in handle for ease of movement
  • 8-degree forward and 3-degree backward locking seat tilt
  • Cast-aluminum base
  • Chrome-hooded, dual-wheel, self-braking, non-marring casters suitable for hard floors
  • Chemical, soil, and puncture resistant polyurethane
  • Class 100/ISO 5 Clean Room
Specifications
Specifications
Standard
Class 100/ISO Class 5
17 in.
No
Ergonomic 8 degree forward and 3 degree backward seat tilt locks in place
1.5 in.
32 lb.
24 in.
Yes
Yes
300 lb.
Cleanroom Industrial Chair
Yes (Pneumatic)
Black
Chrome-hooded dual-wheel, self-braking, non-marring casters (suitable for both hard and carpeted floors)
Polyurethane
Desk Height
15 year parts, lifetime on pneumatic lift
18 in.
No
Yes
17.75 to 23 in.
Polished Cast Aluminum
136 kg