Learn More
Fisherbrand™ Class 100 Newport Industrial Polyurethane Clean Room Chair - Desk Height with Seat Tilt and Casters in Polyurethane Products for Science Education
Fisherbrand™ Newport Class 100 Clean Room Chairs are height adjustable for 31 in. work surfaces and feature ergonomic adjustments, clean room components, and self-braking casters. Self-skinning polyurethane is chemical, soil and puncture resistant and easily cleaned making it ideal for clean room environments.
$296.00
Specifications
Adjustable Yes (Pneumatic)
Material Polyurethane
Footrest No
With Armrest No
With Backrest Yes
View More Specs
Products 1
Catalog Number Color Price Quantity & Availability  
Catalog Number Color Price Quantity & Availability  
S40351 Products for Science Education
View Documents
Black
Each for $296.00
Add to cart
Description
Description
  • Adjustable from 17.75-23 inches from the top of the seat to the floor
  • Features ergonomic adjustments including seat height
  • Locking back tilt
  • Built in handle for ease of movement
  • 8-degree forward and 3-degree backward locking seat tilt
  • Cast-aluminum base
  • Chrome-hooded, dual-wheel, self-braking, non-marring casters suitable for hard floors
  • Chemical, soil, and puncture resistant polyurethane
  • Class 100/ISO 5 Clean Room
Specifications
Specifications
Yes (Pneumatic)
No
Yes
Yes
Polyurethane
No
Yes
Class 100/ISO Class 5