Learn More
Fisherbrand™ Class 100 Polyurethane Clean Room Chair - Desk Height with Medium Back, Seat Tilt, and Stationary Glides in Black Polyurethane
Fisherbrand™ Polyurethane Clean Room Chairs with Medium Back are height adjustable for 31 in. work surfaces and feature ergonomic adjustments and stationary glides. Self-skinning polyurethane is chemical, soil, and puncture resistant and easily cleaned making it ideal for clean room environments.
$432.00 - $438.50
Specifications
Adjustable Yes
Base Material Polished Cast Aluminum
Material Polyurethane
Footrest No
With Armrest No
View More Specs
Products 2
Catalog Number Color Price Quantity & Availability  
Catalog Number Color Price Quantity & Availability  
14-000-714
View Documents
Blue
Each for $432.00
Add to cart
14-400-333
View Documents
Black
Each for $438.50
Add to cart
Description
Description
  • Medium Back
  • Adjustable from 17.5-22.75 inches from the top of the seat to the floor
  • Features ergonomic adjustments including seat height
  • Locking back tilt
  • 8-degree forward and 3-degree backward locking seat tilt
  • Cast-aluminum base
  • Stationary, non-marring glides suitable for hard floors
  • Chemical, soil, and puncture resistant polyurethane
  • Class 100/ISO Class 5 Clean Room
Specifications
Specifications
Yes
Polyurethane
No
No
Desk Height
Polished Cast Aluminum
No
Yes
Yes
Class 100/ISO Class 5