Learn More
Fisherbrand™ Class 100 Polyurethane Clean Room Chair - High Bench Height with Adjustable Arms, Chrome Foot Ring, and Casters in Black Polyurethane
Fisherbrand™ Polyurethane Clean Room Chairs are height adjustable for 42 in. work surfaces & feature ergonomic adjustments, arms, chrome foot ring, & self-braking casters. Self-skinning polyurethane is chemical, soil, & puncture resistant & easily cleaned making it ideal for clean room environments.
$630.50 - $712.00
Specifications
Adjustable Yes
Base Material Polished Cast Aluminum
Material Polyurethane
Footrest Yes
With Armrest Yes
View More Specs
Products 2
Catalog Number Color Price Quantity & Availability  
Catalog Number Color Price Quantity & Availability  
14-000-741
View Documents
Blue
Each for $630.50
Add to cart
14-360-611
View Documents
Black
Each for $712.00
Add to cart
Description
Description
  • Adjustable from 24.5-34.75 inches from the top of the seat to the floor
  • Features ergonomic adjustments including seat height
  • Locking back tilt
  • 6-way adjustable arm rests adjust up and down, in and out, and twist toward and away from the user
  • Chrome adjustable foot ring
  • Cast-aluminum base
  • Chrome-hooded, dual-wheel, self-braking, non-marring casters suitable for hard floors
  • Chemical, soil, and puncture resistant polyurethane
  • Class 100/ISO Class 5 Clean Room
Specifications
Specifications
Yes
Polyurethane
Yes
Yes
High Bench Height
Polished Cast Aluminum
Yes
Yes
No
Class 100/ISO Class 5