Learn More
Fisherbrand™ Class 100 Vinyl Clean Room Chair - Desk Height with Adjustable Arms and Casters in Grade B Vinyl Products for Science Education
Fisherbrand™ Class 100 Vinyl Clean Room Chairs are height adjustable for 31 in. work surfaces and feature ergonomic adjustments, clean room components
$385.00
Specifications
Base Material Polished Cast Aluminum
Arm Style 6-way adjustable up and down, in and out, twist toward or away from the user
Foot Type Chrome-hooded dual-wheel, self-braking, non-marring casters (suitable for both hard and carpeted floors)
Footrest No
Material Vinyl
View More Specs
Products 4
Catalog Number Color Price Quantity & Availability  
Catalog Number Color Price Quantity & Availability  
S0100657 Products for Science Education
View Documents
Orange
Each for $385.00
Add to cart
S0100739 Products for Science Education
View Documents
White
Each for $385.00
Add to cart
S0100740 Products for Science Education
View Documents
Dove
Each for $385.00
Add to cart
S0100741 Products for Science Education
View Documents
Taupe
Each for $385.00
Add to cart
Description
Description
  • Adjustable from 19 to 24 inches from the top of the seat to the floor
  • Features ergonomic adjustments including seat height
  • Locking back tilt
  • 6-way adjustable arm rests adjust up and down, in and out, and twist toward and away from the user
  • Cast-aluminum base
  • Chrome-hooded, dual-wheel, self-braking, non-marring casters suitable for hard floors
  • Antibacterial/antifungal Grade B Vinyl
  • Variety of color options to suit any work space
  • Class 100/ISO 5 Clean Room
Specifications
Specifications
Polished Cast Aluminum
Chrome-hooded dual-wheel, self-braking, non-marring casters (suitable for both hard and carpeted floors)
Vinyl
Desk Height
Yes
No
6-way adjustable up and down, in and out, twist toward or away from the user
No
Cleanroom Seating
41 lb.
Yes