Learn More
Fisherbrand™ Class 100 Vinyl Clean Room Chair - Desk Height with Adjustable Arms and Casters in Grade B Vinyl
Fisherbrand™ Class 100 Vinyl Clean Room Chairs are height adjustable for 31 in. work surfaces and feature ergonomic adjustments, clean room components, adjustable arms, & self-braking casters. Antibacterial/antimicrobial vinyl is easily cleaned making it ideal for clean room environments.
$515.50
Specifications
Adjustable Yes
Base Material Polished Cast Aluminum
Material Vinyl
Footrest No
With Armrest Yes
View More Specs
Products 5
Catalog Number Color Price Quantity & Availability  
Catalog Number Color Price Quantity & Availability  
14-360-504
View Documents
Blue
Each for $515.50
Add to cart
14-360-506
View Documents
Black
Each for $515.50
Add to cart
14-360-507
View Documents
Burgundy
Each for $515.50
Add to cart
14-360-505
View Documents
Gray
Each for $515.50
Add to cart
14-360-508
View Documents
Dark Blue
Each for $515.50
Add to cart
Description
Description
  • Adjustable from 19-24 inches from the top of the seat to the floor
  • Features ergonomic adjustments including seat height
  • Locking back tilt
  • 6-way adjustable arm rests adjust up and down, in and out, and twist toward and away from the user
  • Cast-aluminum base
  • Chrome-hooded, dual-wheel, self-braking, non-marring casters suitable for hard floors
  • Antibacterial/antifungal Grade B Vinyl
  • Class 100/ISO Class 5 Clean Room
Specifications
Specifications
Yes
Vinyl
Yes
Yes
Desk Height
Polished Cast Aluminum
No
Yes
No
Class 100/ISO Class 5