Learn More
Fisherbrand Class 100 Vinyl Clean Room Chair - Desk Height with Casters in Grade B Vinyl
Fisherbrand Class 100 Vinyl Clean Room Chairs are height adjustable for 31 in. work surfaces and feature ergonomic adjustments, clean room components, and self-braking casters. Antibacterial/antimicrobial vinyl is easily cleaned making it ideal for clean room environments.
$292.00
Specifications
Adjustable | Yes |
---|---|
Base Style | Polished Cast Aluminum |
Material | Trailblazer Grade B Vinyl |
Footrest | No |
With Armrest | No |
Description
- Adjustable from 19-24 inches from the top of the seat to the floor
- Features ergonomic adjustments including seat height
- Locking back tilt
- Cast-aluminum base
- Chrome-hooded, dual-wheel, self-braking, non-marring casters suitable for hard floors
- Antibacterial/antifungal Grade B Vinyl
- Variety of color options to suit any work space
- Class 100/ISO Class 5 Clean Room
Specifications
Yes | |
Trailblazer Grade B Vinyl | |
No | |
Yes | |
Desk Height |
Polished Cast Aluminum | |
No | |
Yes | |
No | |
Class 100/ISO Class 5 |