Learn More
Fisherbrand™ Class 100 Vinyl Clean Room Chair - Desk Height with Seat Tilt Products for Science Education
Fisherbrand™ Class 100 Vinyl Clean Room Chairs are height adjustable for 31 in. work surfaces and feature ergonomic adjustments, clean room components
$417.00
Specifications
Base Material Polished Cast Aluminum
Arm Style 6-way adjustable up and down, in and out, twist toward or away from the user
Foot Type Chrome-hooded dual-wheel, self-braking, non-marring casters (suitable for both hard and carpeted floors)
Footrest No
Includes Round tube chrome foot ring with internal lock
View More Specs
Products 4
Catalog Number Color Price Quantity & Availability  
Catalog Number Color Price Quantity & Availability  
S0100617 Products for Science Education
View Documents
White
Each for $417.00
Add to cart
S0100618 Products for Science Education
View Documents
Dove
Each for $417.00
Add to cart
S0100619 Products for Science Education
View Documents
Taupe
Each for $417.00
Add to cart
S0100620 Products for Science Education
View Documents
Orange
Each for $417.00
Add to cart
Description
Description
  • Adjustable from 19 to 24 inches from the top of the seat to the floor
  • Features ergonomic adjustments including seat height
  • Locking back tilt
  • 8-degree forward and 3-degree backward locking seat tilt
  • 6-way adjustable arm rests adjust up and down, in and out, and twist toward and away from the user
  • Cast-aluminum base
  • Chrome-hooded, dual-wheel, self-braking, non-marring casters suitable for hard floors
  • Antibacterial/antifungal Grade B Vinyl
  • Variety of color options to suit any work space
  • Class 100/ISO 5 Clean Room
Specifications
Specifications
Polished Cast Aluminum
Chrome-hooded dual-wheel, self-braking, non-marring casters (suitable for both hard and carpeted floors)
Round tube chrome foot ring with internal lock
Cleanroom Seating
Desk Height
Yes
Yes
6-way adjustable up and down, in and out, twist toward or away from the user
No
Vinyl
Ergonomic 8 degree forward and 3 degree backward seat tilt locks in place
42 lb.
Yes