Learn More
Fisherbrand™ Class 100 Vinyl Clean Room Chair - High Bench Height with Adj Arms, Chrome Foot Ring, and Stationary Glides in Grade B Vinyl
SDPSCI_ED
Fisherbrand™ Class 100 Vinyl Clean Room Chairs are height adjustable for 42 in. work surfaces and feature ergonomic adjustments, clean room components, foot ring, arms, and stationary glides. Antibacterial/Antifungal Grade B vinyl is easily cleaned making it ideal for clean room environments.
Specifications
Adjustable Yes
Adjustable (Height) 26.25 to 36 in.
Arm Style 6-way adjustable (move up and down, in and out, and twist toward or away from the user)
Back Style Standard
Base Material Polished Cast Aluminum
View More Specs
Products 5
Catalog Number Color Price Quantity & Availability  
Catalog Number Color Price Quantity & Availability  
S36651
SDPSCI_ED
View Documents
Blue Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area.
View alternate product
N/A
S38467
SDPSCI_ED
View Documents
Black Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area.
View alternate product
N/A
S38660
SDPSCI_ED
View Documents
Burgundy Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area.
View alternate product
N/A
S38466
SDPSCI_ED
View Documents
Gray Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area.
View alternate product
N/A
S38661
SDPSCI_ED
View Documents
Dark Blue Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area.
View alternate product
N/A
Description
Description
  • Adjustable from 26.25-36 inches from the top of the seat to the floor
  • Features ergonomic adjustments including seat height
  • Locking back tilt
  • 6-way adjustable arm rests adjust up and down, in and out, and twist toward and away from the user
  • 20-Inch Chrome adjustable foot ring with internal lock
  • Cast-aluminum base
  • Stationary, non-marring glides suitable for hard floors
  • Antibacterial/antifungal Grade B Vinyl
  • Variety of color options to suit any work space
  • Class 100/ISO 5 Clean Room
Specifications
Specifications
Yes
6-way adjustable (move up and down, in and out, and twist toward or away from the user)
Polished Cast Aluminum
20 in.
Round tube chrome foot ring with internal lock
136 kg
46 lb.
Yes
Yes
Cleanroom Chair
26.25 to 36 in.
Standard
Class 100/ISO Class 5
Black-non-marring bell glides (suitable for both hard and carpeted floors)
300 lb.
High Bench Height
Yes
No
No
Vinyl