Learn More
Fisherbrand™ Class 100 Vinyl Clean Room Chair - High Bench Height with Adjustable Arms, Chrome Foot Ring, and Stationary Glides in Grade A Vinyl Products for Science Education
Fisherbrand™ Class 100 Vinyl Clean Room Chairs are height adjustable for 42 in. work surfaces and feature ergonomic adjustments, clean room components, foot ring, arms, and stationary glides. Grade A vinyl, in a variety of colors, offer additional antibacterial properties and a high-end look and feel.
$475.00
Specifications
Base Material Polished Cast Aluminum
Arm Style 6-way adjustable up and down, in and out, twist toward or away from the user
Foot Type Non-marring bell glides (suitable for both hard and carpeted floors)
Footrest Yes
Footrest Size (English) 20 in.
View More Specs
Products 6
Catalog Number Color Price Quantity & Availability  
Catalog Number Color Price Quantity & Availability  
S0100255 Products for Science Education
View Documents
Marine Blue
Each for $475.00
Add to cart
S0100310 Products for Science Education
View Documents
Taupe
Each for $475.00
Add to cart
S0100391 Products for Science Education
View Documents
Sterling
Each for $475.00
Add to cart
S0100392 Products for Science Education
View Documents
Carbon
Each for $475.00
Add to cart
S0100393 Products for Science Education
View Documents
Storm
Each for $475.00
Add to cart
S0100394 Products for Science Education
View Documents
Borscht
Each for $475.00
Add to cart
Description
Description
  • Adjustable from 26.25 to 36 inches from the top of the seat to the floor
  • Features ergonomic adjustments including seat height
  • Locking back tilt
  • 6-way adjustable arm rests adjust up and down, in and out, and twist toward and away from the user
  • 20-Inch Chrome adjustable foot ring with internal lock
  • Cast-aluminum base
  • Stationary, non-marring glides suitable for hard floors
  • Antibacterial/antifungal Grade A Vinyl
  • Additional antibacterial protections
  • High end look and feel
  • Variety of color options to suit any work space
  • Class 100/ISO Class 5 Clean Room
Specifications
Specifications
Polished Cast Aluminum
Non-marring bell glides (suitable for both hard and carpeted floors)
20 in.
Cleanroom Seating
46 lb.
No
6-way adjustable up and down, in and out, twist toward or away from the user
Yes
Vinyl
High Bench Height
Yes
No