Learn More
Fisherbrand™ Class 100 Vinyl Clean Room Chair - High Bench Height with Chrome Foot Ring and Stationary Glides in Grade B Vinyl
Fisherbrand™ Class 100 Vinyl Clean Room Chairs are height adjustable for 42 in. work surfaces and feature ergonomic adjustments, clean room components, foot ring, and stationary glides. Antibacterial/Antifungal Grade B vinyl is easily cleaned making it ideal for clean room environments.
$491.00
Specifications
Adjustable Yes
Base Material Polished Cast Aluminum
Material Vinyl
Footrest Yes
With Armrest No
View More Specs
Products 5
Catalog Number Color Price Quantity & Availability  
Catalog Number Color Price Quantity & Availability  
14-360-579
View Documents
Blue
Each for $491.00
Add to cart
14-360-581
View Documents
Black
Each for $491.00
Add to cart
14-360-582
View Documents
Burgundy
Each for $491.00
Add to cart
14-360-580
View Documents
Gray
Each for $491.00
Add to cart
14-360-583
View Documents
Dark Blue
Each for $491.00
Add to cart
Description
Description
  • Adjustable from 26.25-36 inches from the top of the seat to the floor
  • Features ergonomic adjustments including seat height
  • Locking back tilt
  • 20-Inch Chrome adjustable foot ring with internal lock
  • Cast-aluminum base
  • Stationary, non-marring glides suitable for hard floors
  • Antibacterial/antifungal Grade B Vinyl
  • Variety of color options to suit any work space
  • Class 100/ISO Class 5 Clean Room
Specifications
Specifications
Yes
Vinyl
No
No
High Bench Height
Polished Cast Aluminum
Yes
Yes
No
Class 100/ISO Class 5