Learn More
Fisherbrand™ Class 100 Vinyl Clean Room Chair - High Bench Height with Medium Back, Chrome Foot Ring, and Casters in Grade B Vinyl Products for Science Education
Fisherbrand™ Class 100 Vinyl Clean Room Chairs with Medium Back are height adjustable for 42 in. work surfaces and feature ergonomic adjustments, foot ring, and self-braking casters. Antibacterial/Antifungal Grade B vinyl is easily cleaned making them ideal for clean room environments.
$420.00
Specifications
Adjustable Yes
Base Material Polished Cast Aluminum
Material Vinyl
Footrest Yes
With Armrest No
View More Specs
Products 4
Catalog Number Color Price Quantity & Availability  
Catalog Number Color Price Quantity & Availability  
S0101042 Products for Science Education
View Documents
Taupe
Each for $420.00
Add to cart
S0101103 Products for Science Education
View Documents
Orange
Each for $420.00
Add to cart
S0101113 Products for Science Education
View Documents
Dove
Each for $420.00
Add to cart
S0101123 Products for Science Education
View Documents
White
Each for $420.00
Add to cart
Description
Description
  • Medium Back
  • Adjustable from 26.25-36 inches from the top of the seat to the floor
  • Features ergonomic adjustments including seat height
  • Locking back tilt
  • 20-Inch Chrome adjustable foot ring with internal lock
  • Cast-aluminum base
  • Chrome-hooded, dual-wheel, self-braking, non-marring casters suitable for hard floors
  • Antibacterial/antifungal Grade B Vinyl
  • Variety of color options to suit any work space
  • Class 100/ISO Class 5 Clean Room
Specifications
Specifications
Yes
Vinyl
No
Yes
High Bench Height
Polished Cast Aluminum
Yes
Yes
No
Class 100/ISO Class 5