Learn More
Fisherbrand™ Class 100 Vinyl Clean Room Chair - High Bench Height with Medium Back, Chrome Foot Ring, and Stationary Glides in Grade B Vinyl
Fisherbrand™ Class 100 Vinyl Clean Room Chairs with Medium Back are height adjustable for 42 in. work surfaces and feature ergonomic adjustments, foot ring, and stationary glides. Antibacterial/Antifungal Grade B vinyl is easily cleaned making them ideal for clean room environments.
$846.33
Specifications
Base Style Polished Cast Aluminum Base
Foot Type Black-non-marring bell glides (suitable for both hard and carpeted floors)
Footrest Yes
Footrest Size (English) 20 in.
Material Trailblazer Grade B Vinyl
View More Specs
Products 4
Catalog Number Color Price Quantity & Availability  
Catalog Number Color Price Quantity & Availability  
15-002-365
View Documents
Adobe White
Each for $846.33
Add to cart
15-002-366
View Documents
Dove
Each for $846.33
Add to cart
15-002-367
View Documents
Taupe
Each for $846.33
Add to cart
15-002-368
View Documents
Orange Kist
Each for $846.33
Add to cart
Description
Description
  • Medium Back
  • Adjustable from 26.25 to 36 inches from the top of the seat to the floor
  • Features ergonomic adjustments including seat height
  • Locking back tilt
  • 20-Inch Chrome adjustable foot ring with internal lock
  • Cast-aluminum base
  • Stationary, non-marring glides suitable for hard floors
  • Antibacterial/antifungal Grade B Vinyl
  • Variety of color options to suit any work space
  • Class 100/ISO Class 5 Clean Room
Specifications
Specifications
Polished Cast Aluminum Base
Yes
Trailblazer Grade B Vinyl
No
No
Black-non-marring bell glides (suitable for both hard and carpeted floors)
20 in.
37 lb.
No