Learn More
Fisherbrand™ Class 100 Vinyl Clean Room Chair - High Bench Height with Seat Tilt, Adj Arms, Foot Ring, and Stationary Glides in Grade B Vinyl
Fisherbrand™ Class 100 Vinyl Clean Room Chairs are height adjustable for 42 in. work surfaces and feature ergonomic adjustments, clean room components, foot ring, arms, and stationary glides. Antibacterial/Antifungal Grade B vinyl is easily cleaned making it ideal for clean room environments.
$655.00
Specifications
Adjustable Yes
Base Material Polished Cast Aluminum
Material Vinyl
Footrest Yes
With Armrest Yes
View More Specs
Products 5
Catalog Number Color Price Quantity & Availability  
Catalog Number Color Price Quantity & Availability  
14-360-594
View Documents
Blue
Each for $655.00
Add to cart
14-360-596
View Documents
Black
Each for $655.00
Add to cart
14-360-597
View Documents
Burgundy
Each for $655.00
Add to cart
14-360-595
View Documents
Gray
Each for $655.00
Add to cart
14-360-598
View Documents
Dark Blue
Each for $655.00
Add to cart
Description
Description
  • Adjustable from 26.25-36 inches from the top of the seat to the floor
  • Features ergonomic adjustments including seat height
  • Locking back tilt
  • 8-degree forward and 3-degree backward locking seat tilt
  • 6-way adjustable arm rests adjust up and down, in and out, and twist toward and away from the user
  • 20-Inch Chrome adjustable foot ring with internal lock
  • Cast-aluminum base
  • Stationary, non-marring glides suitable for hard floors
  • Antibacterial/antifungal Grade B Vinyl
  • Variety of color options to suit any work space
  • Class 100/ISO 5 Clean Room
Specifications
Specifications
Yes
Vinyl
Yes
No
High Bench Height
Polished Cast Aluminum
Yes
Yes
Yes
Class 100/ISO Class 5