Learn More
Fisherbrand™ Class 100 Vinyl Clean Room Chair - Medium Bench Height with Adj Arms, Chrome Foot Ring, and Stationary Glides in Grade B Vinyl
SCI_ED
Fisherbrand™ Class 100 Vinyl Clean Room Chairs are height adjustable for 36 in. work surfaces and feature ergonomic adjustments, clean room components, foot ring, arms, and stationary glides. Antibacterial/Antifungal Grade B vinyl is easily cleaned making it ideal for clean room environments.
 View more versions of this product
Catalog No. S36739
Add to cart
Description
Description
  • Adjustable from 21.5-29 inches from the top of the seat to the floor
  • Features ergonomic adjustments including seat height
  • Locking back tilt
  • 6-way adjustable arm rests adjust up and down, in and out, and twist toward and away from the user
  • 20-Inch Chrome adjustable foot ring with internal lock
  • Cast-aluminum base
  • Stationary, non-marring glides suitable for hard floors
  • Antibacterial/antifungal Grade B Vinyl
  • Variety of color options to suit any work space
  • Class 100/ISO 5 Clean Room
Specifications
Specifications
Standard
6-way adjustable (move up and down, in and out, and twist toward or away from the user)
Burgundy
Yes
Round tube chrome foot ring with internal lock
Medium Bench Height
Yes
No
21.5 to 29 in.
Polished Cast Aluminum
136 kg
Yes
Class 100/ISO Class 5
Black-non-marring bell glides (suitable for both hard and carpeted floors)
20 in.
Vinyl
44 lb.
Yes
No
300 lb.
Cleanroom Chair