Learn More
Fisherbrand™ Class 100 Vinyl Clean Room Chair - Medium Bench Height with Medium Back, Adj Arms, Foot Ring, and Casters in Grade B Vinyl Products for Science Education
Fisherbrand™ Class 100 Vinyl Clean Room Chairs with Medium Back are height adjustable for 36 in. work surfaces and feature ergonomic adjustments, clean room components, arms, & self-braking casters. Antibacterial/Antifungal Grade B vinyl is easily cleaned making ideal for clean room environments.
$392.00
Specifications
Adjustable Yes
Material Vinyl
Footrest Yes
With Armrest Yes
With Backrest Yes
View More Specs
Products 5
Catalog Number Color Price Quantity & Availability  
Catalog Number Color Price Quantity & Availability  
S40032 Products for Science Education
View Documents
Blue
Each for $392.00
Add to cart
S40062 Products for Science Education
View Documents
Black
Each for $392.00
Add to cart
S40087 Products for Science Education
View Documents
Burgundy
Each for $392.00
Add to cart
S40097 Products for Science Education
View Documents
Gray
Each for $392.00
Add to cart
S40147 Products for Science Education
View Documents
Dark Blue
Each for $392.00
Add to cart
Description
Description
  • Medium Back
  • Adjustable from 21.5-29 inches from the top of the seat to the floor
  • Features ergonomic adjustments including seat height
  • Locking back tilt
  • 6-way adjustable arm rests adjust up and down, in and out, and twist toward and away from the user
  • Chrome adjustable foot ring
  • Cast-aluminum base
  • Chrome-hooded, dual-wheel, self-braking, non-marring casters suitable for hard floors
  • Antibacterial/antifungal Grade B Vinyl
  • Class 100/ISO Class 5 Clean Room
Specifications
Specifications
Yes
Yes
Yes
No
Vinyl
Yes
Yes
Class 100/ISO Class 5