Learn More
Fisherbrand™ Class 100 Vinyl Clean Room/ESD Chair - Desk Height with Adjustable Arms and ESD Casters in ESD Vinyl
SCI_ED
Fisherbrand™ Class 100 Vinyl Clean Room/ESD Chairs are height adjustable for 31 in. work surfaces and feature ergonomic adjustments, clean room and electro-static dissipating components, adjustable arms, & self-braking casters. Properly grounded for static sensitive applications.
 View more versions of this product
Catalog No. S36823
Add to cart
Description
Description
  • Adjustable from 19-24 inches from the top of the seat to the floor
  • Features ergonomic adjustments including seat height
  • Locking back tilt
  • 6-way adjustable arm rests adjust up and down, in and out, and twist toward and away from the user
  • Electro-static dissipating materials for proper grounding
  • Cast-aluminum base
  • Dual-wheel, self-braking, non-marring ESD casters suitable for hard floors
  • Electro-static dissipating vinyl
  • Meets ANSI/ESD STM12.1-2013
  • Class 100/ISO Class 5 Clean Room
Specifications
Specifications
Standard
6-way adjustable (move up and down, in and out, and twist toward or away from the user)
Black
No
Desk Height
Yes
Yes
19 to 24 in.
Polished Cast Aluminum
136 kg
Yes
Class 100/ISO Class 5
Electro-Static Dissipating, dual-wheel, non-marring casters (suitable for both hard and carpeted floors)
ESD Upholstery
41 lb.
Yes
No
300 lb.
Cleanroom ESD Chair