Learn More
Fisherbrand™ Class 100 Vinyl Clean Room/ESD Chair - High Bench Height with Adj Arms, Chrome Foot Ring, and ESD Stationary Glides in ESD Vinyl
SCI_ED
Fisherbrand™ Class 100 Vinyl Clean Room/ESD Chairs are height adjustable for 42 in. work surfaces and feature ergonomic adjustments, clean room and electro-static dissipating components, adjustable arms, chrome adjustable foot ring, and stationary glides. Properly grounded for static sensitive applications.
 View more versions of this product
Catalog No. S36970
Add to cart
Description
Description
  • Adjustable from 26.25-36 inches from the top of the seat to the floor
  • Features ergonomic adjustments including seat height
  • Locking back tilt
  • 6-way adjustable arm rests adjust up and down, in and out, and twist toward and away from the user
  • Electro-static dissipating components to ensure proper grounding
  • 20-Inch Chrome adjustable foot ring with internal lock
  • Cast-aluminum base
  • Stationary, non-marring ESD glides suitable for hard floors
  • Electro-static dissipating vinyl
  • Meets ANSI/ESD STM12.1-2013
  • Class 100/ISO Class 5 Clean Room
Specifications
Specifications
Standard
6-way adjustable (move up and down, in and out, and twist toward or away from the user)
Black
Yes
ESD Upholstery
46 lb.
Yes
No
300 lb.
Cleanroom ESD Chair
Yes
Class 100/ISO Class 5
Electro-Static Dissipating, non-marring bell glides (suitable for both hard and carpeted floors)
Round tube chrome foot ring with internal lock
High Bench Height
Yes
No
26.25 to 36 in.
Polished Cast Aluminum
136 kg