Learn More
Fisherbrand™ Class 100 Vinyl Clean Room/ESD Chair - Med Bench Height with Medium Back, Adj Arms, Foot Ring, and ESD Stationary Glides in ESD Vinyl
SCI_ED
Fisherbrand™ Class 100 Vinyl Clean Room/ESD Chairs with Medium Back height adjustable for 36 in. work surfaces & feature ergonomic adjustments, clean room & electro-static dissipating components, arms, chrome adjustable foot ring, & stationary glides. Properly grounded for static sensitive applications.
 View more versions of this product
Catalog No. S39965
Add to cart
Description
Description
  • Medium Back
  • Adjustable from 21.5-29 inches from the top of the seat to the floor
  • Features ergonomic adjustments including seat height
  • Locking back tilt
  • 6-way adjustable arm rests adjust up and down, in and out, and twist toward and away from the user
  • Chrome adjustable foot ring
  • Electro-static dissipating materials for proper grounding
  • Cast-aluminum base
  • Stationary ESD glides suitable for hard floors
  • Electro-static dissipating vinyl
  • Meets ANSI/ESD STM12.1-2013
  • Class 100/ISO Class 5 Clean Room
Specifications
Specifications
Standard
6-way adjustable (move up and down, in and out, and twist toward or away from the user)
Black
Yes
Round tube chrome foot ring with internal lock
Medium Bench Height
Yes
No
21.5 to 29 in.
Polished Cast Aluminum
136 kg
Yes
Class 100/ISO Class 5
Electro-Static Dissipating, non-marring bell glides (suitable for both hard and carpeted floors)
20 in.
ESD Upholstery
45 lb.
Yes
No
300 lb.
Cleanroom ESD Chair