Learn More
Fisherbrand™ Class 100 Vinyl Clean Room/ESD Chair - Medium Bench Height with Seat Tilt, Adj Arms, Foot Ring, and ESD Stationary Glides in ESD Vinyl
SDPSCI_ED
Fisherbrand™ Class 100 Vinyl Clean Room/ESD Chairs are height adjustable for 36 in. work surfaces & feature ergonomic adjustments, clean room & electro-static dissipating components, adjustable arms, chrome adjustable foot ring, & stationary glides. Properly grounded for static sensitive applications.
 View more versions of this product
Catalog No. S36257
Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area.
View alternate product
Description
Description
  • Adjustable from 21.5-29 inches from the top of the seat to the floor
  • Features ergonomic adjustments including seat height
  • Locking back tilt
  • Locking seat tilt
  • 6-way adjustable arm rests adjust up and down, in and out, and twist toward and away from the user
  • Chrome adjustable foot ring
  • Electro-static dissipating materials for proper grounding
  • Cast-aluminum base
  • Stationary ESD glides suitable for hard floors
  • Electro-static dissipating vinyl
  • Meets ANSI/ESD STM12.1-2013
  • Class 100/ISO Class 5 Clean Room
Specifications
Specifications
Cleanroom ESD Chair
Standard
Yes
Yes
N/A
Yes
Ergonomic 8 degree forward and 3 degree backward seat tilt locks in place
No
21.5 to 29 in.
Polished Cast Aluminum
Class 100/ISO Class 5
Yes
Medium Bench Height
ESD Upholstery
Electro-Static Dissipating, non-marring bell glides (suitable for both hard and carpeted floors)
6-way adjustable (move up and down, in and out, and twist toward or away from the user)
45 lb.
Blue
Round tube chrome foot ring with internal lock
Product Suggestions
Product Suggestions
Safety and Handling
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov