Learn More
Fisherbrand™ Class 100 Vinyl CR Chair - High Bench Height with Med Back, Seat Tilt, Foot Ring, and Stationary Glides in Grade B Vinyl
Fisherbrand™ Class 100 Vinyl Clean Room Chairs with Medium Back are height adjustable for 42 in. work surfaces and feature ergonomic adjustments, foot ring, and stationary glides. Antibacterial/Antifungal Grade B vinyl is easily cleaned making them ideal for clean room environments.
$505.50
Specifications
Adjustable Yes
Material Vinyl
Footrest Yes
With Armrest No
With Backrest Yes
View More Specs
Products 5
Catalog Number Color Price Quantity & Availability  
Catalog Number Color Price Quantity & Availability  
14-000-389
View Documents
Black
Each for $505.50
Add to cart
14-000-391
View Documents
Blue
Each for $505.50
Add to cart
14-000-399
View Documents
Burgundy
Each for $505.50
Add to cart
14-000-407
View Documents
Gray
Each for $505.50
Add to cart
14-000-415
View Documents
Dark Blue
Each for $505.50
Add to cart
Description
Description
  • Medium Back
  • Adjustable from 26.25-36 inches from the top of the seat to the floor
  • Features ergonomic adjustments including seat height
  • Locking back tilt
  • 8-degree forward and 3-degree backward locking seat tilt
  • 20-Inch Chrome adjustable foot ring with internal lock
  • Cast-aluminum base
  • Stationary, non-marring glides suitable for hard floors
  • Antibacterial/antifungal Grade B Vinyl
  • Variety of color options to suit any work space
  • Class 100/ISO 5 Clean Room
Specifications
Specifications
Yes
Yes
Yes
Yes
Vinyl
No
No
Class 100/ISO Class 5