Learn More
Fisherbrand™ Class 100 Vinyl CR Chair - High Bench Height with Med Back, Tilt, Adj Arms, Foot Ring, and Stationary Glides in Grade B Vinyl
SDPSCI_ED
Fisherbrand™ Class 100 Vinyl Clean Room Chairs with Medium Back are height adjustable for 42 in. work surfaces and feature ergonomic adjustments, foot ring, arms, and stationary glides. Antibacterial/Antifungal Grade B vinyl is easily cleaned making them ideal for clean room environments.
 View more versions of this product
Catalog No. S40231
Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area.
View alternate product
Description
Description
  • Medium Back
  • Adjustable from 26.25-36 inches from the top of the seat to the floor
  • Features ergonomic adjustments including seat height
  • Locking back tilt
  • 8-degree forward and 3-degree backward locking seat tilt
  • 6-way adjustable arm rests adjust up and down, in and out, and twist toward and away from the user
  • 20-Inch Chrome adjustable foot ring with internal lock
  • Cast-aluminum base
  • Stationary, non-marring glides suitable for hard floors
  • Antibacterial/antifungal Grade B Vinyl
  • Variety of color options to suit any work space
  • Class 100/ISO 5 Clean Room
Specifications
Specifications
Cleanroom Chair
Yes
Ergonomic 8 degree forward and 3 degree backward seat tilt locks in place
48 lb.
Yes
Round tube chrome foot ring with internal lock
Class 100/ISO Class 5
20 in.
26.25 to 36 in.
6-way adjustable (move up and down, in and out, and twist toward or away from the user)
Polished Cast Aluminum
No
Yes
High Bench Height
136 kg
Yes
300 lb.
Black-non-marring bell glides (suitable for both hard and carpeted floors)
Dark Blue
Yes
Standard
Vinyl
Product Suggestions
Product Suggestions
Safety and Handling
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov